Learn More
MSC Milwaukee Tool Hedge Trimmer, 18V, 30 in Blade, Double-Sided
Supplier: MSC 51885069
The M18 FUEL 30 inch Hedge Trimmer is designed for landscape maintenance professionals, delivering power, performance, and durability. Its POWERSTATE Brushless Motor cuts branches up to 1-1/4 inches, facilitating aggressive cut-backs and faster trimming. Features include a rotating handle, double-sided blades, and a clippings sweep attachment for enhanced control and clean cuts. REDLINK PLUS Intelligence ensures optimized performance and productivity across the M18 System.
- Power to cut 1-1/4 inch branches for demanding cutbacks and faster trimming.
- Rotating handle and double-sided blades provide improved control and clean cuts.
- 30 inch dual-sharpened blade set with 1-1/4 inch blade gap.
- 3,200 strokes per minute for efficient trimming.
- Blade tip guard prevents damage to property and protects blades.
- Cordless and battery powered.