Learn More
ILEX LIFE SCIENCES LLC M199 EBSS P04-07500 500 ML
SDP
Catalog No. 502733862
Encompass_Preferred
PAN-Biotech Medium 199 (M199) w: EBSS, w/o: L-Glutamine, w: 2.2 g/L NaHCO3 (500 ml). EBSS: Contains Earle′s Balanced Salt Solution (EBSS). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Medium 199, also known as M199, was originally developed to assay the nutrient demand of embryonic chicken fibroblasts. It also works very well with cells from many different animal species. Serum should be added for long term cultures.
Catalog No. 50-273-3862 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0407500
Add to Cart
Add to Cart
PAN-Biotech Medium 199 (M199) w: EBSS, w/o: L-Glutamine, w: 2.2 g/L NaHCO3 (500 ml). EBSS: Contains Earle′s Balanced Salt Solution (EBSS). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Medium 199, also known as M199, was originally developed to assay the nutrient demand of embryonic chicken fibroblasts. It also works very well with cells from many different animal species. Serum should be added for long term cultures.