Learn More
TRIBIOSCIENCE INC M2 Mouse Embryo Medium, 50 ML
SDP
Catalog No. NC3454603
Encompass_Preferred
Tribioscience’s Mouse M2 Medium is a chemically-defined medium for culturing preimplantation stage mouse embryos. It is used for collecting and handling mouse embryos for prolonged periods in a CO2 incubator.
Catalog No. NC3454603 Supplier TRIBIOSCIENCE INC Supplier No. TBS8070
Tribioscience’s Mouse M2 Medium is a chemically-defined medium for culturing preimplantation stage mouse embryos. It is used for collecting and handling mouse embryos for prolonged periods in a CO2 incubator.