Learn More
Alkali Scientific Alkali Scientific™ M2100 TubeRocker™ for Laboratory Tubes
SDP
Catalog No. 503029591
Encompass_Preferred
Ideal for tube sizes 1.5ml to 50ml Optomized speed and tilt angle for thorough mixing in sample tubes Cold room and incubator safe No adapters or mounting hardware required The Alkali Scientific™ M2100 TubeRocker Mini is designed to provide the ideal speed and tilt angle (24 rpm / 30 °) for agitation of sample tubes. The grooved design of the included rubber mat properly secures all tube sizes ranging from 1.5 to 50ml,without the use of added straps or bands. There are no complicated controls, no settings or adjustments to be made; just press the power switch and let it rock. Small in size (only 5 x 13 inches) and able to with stand temperatures from 4ºC to 65ºC, the TubeRocker Mini is small enough to fit on any lab bench and can conveniently be placed inside a cold room, refrigerator or laboratory incubator.  
Catalog No. 50-302-9591 Supplier Alkali Scientific Supplier No. RS7004
Ideal for tube sizes 1.5ml to 50ml Optomized speed and tilt angle for thorough mixing in sample tubes Cold room and incubator safe No adapters or mounting hardware required The Alkali Scientific™ M2100 TubeRocker Mini is designed to provide the ideal speed and tilt angle (24 rpm / 30 °) for agitation of sample tubes. The grooved design of the included rubber mat properly secures all tube sizes ranging from 1.5 to 50ml,without the use of added straps or bands. There are no complicated controls, no settings or adjustments to be made; just press the power switch and let it rock. Small in size (only 5 x 13 inches) and able to with stand temperatures from 4ºC to 65ºC, the TubeRocker Mini is small enough to fit on any lab bench and can conveniently be placed inside a cold room, refrigerator or laboratory incubator.  