Learn More
Covaris Inc M220 Holder XTU Insert microTUBE 15 µl
SDP
Catalog No. NC3452940
Encompass
This is an insert specifically designed to hold a microTUBE 15 µl in a M220 Holder XTU. It requires M220 Holder XTU (product 500414). For use with microTUBE-15 AFA Beads Screw-Cap (product 520145).
Catalog No. NC3452940 Supplier Covaris Inc Supplier No. 500420
This is an insert specifically designed to hold a microTUBE 15 µl in a M220 Holder XTU. It requires M220 Holder XTU (product 500414). For use with microTUBE-15 AFA Beads Screw-Cap (product 520145).