Learn More
Markson Labsales M261 COLOR YELLOW PH7 BUFFER
SDP
Supplier:  Markson Labsales M261
Encompass_Preferred
Markson NBS Traceable Buffers, NBS Traceable Buffers High Precision pH Standards,500 mL pH 7,Yellow,High Precision pH Standards + +/-0.002 accuracy!,These NBS traceable standards are supplied in special pint polyester bo
Catalog No. NC9773180