Learn More
TARGETMOL CHEMICALS INC M2698 5MG
SDP
Catalog No. 502581500
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. M2698 (MSC2363318A) is an inhibitor of p70S6K Akt1 and Akt3 with IC50s of 1 nM. M2698 shows anti-cancer activity. purity: 99%
Catalog No. 50-258-1500 Supplier TARGETMOL CHEMICALS INC Supplier No. T119285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. M2698 (MSC2363318A) is an inhibitor of p70S6K Akt1 and Akt3 with IC50s of 1 nM. M2698 shows anti-cancer activity. purity: 99%