Learn More
Chemglass Life Sciences M27 Blind Stopper for Bioreactor Head Plate (Chemglass CLS-1385-14)
SDP
Catalog No. 502733307
Encompass
The M27 Blind Stopper is a threaded blind plug designed to close unused M27 threaded ports on bioreactor head plates. It provides a corrosion-resistant closure for unused openings and is suitable as a replacement component in laboratory bioreactor assemblies.
  • Constructed from 316L stainless steel.
  • Electropolished surface finish of 20Ra or better.
  • Threads into M27 ports from the outside to close unused openings.
  • Designed for use with bioreactor head plates and assemblies.
  • Compatible with common bioreactor systems; replacement silicone o-ring available.
Catalog No. 50-273-3307 Supplier Chemglass Life Sciences Supplier No. CLS138514
The M27 Blind Stopper is a threaded blind plug designed to close unused M27 threaded ports on bioreactor head plates. It provides a corrosion-resistant closure for unused openings and is suitable as a replacement component in laboratory bioreactor assemblies.
  • Constructed from 316L stainless steel.
  • Electropolished surface finish of 20Ra or better.
  • Threads into M27 ports from the outside to close unused openings.
  • Designed for use with bioreactor head plates and assemblies.
  • Compatible with common bioreactor systems; replacement silicone o-ring available.