Learn More
Hach Company Radiometer Analytical M295Ag Metal Electrode (silver rod sensor, PTFE body, banana plug)
Catalog No. 501997601 Shop All Hach Company Products
Encompass
The Radiometer M295Ag Metal Electrode has a PTFE body and silver rod sensor. It is recommended for silver halide titrations. The M295Ag has a fixed 1 meter cable with banana plug.
Catalog No. 50-199-7601 Supplier Hach Company Supplier No. E34M003
The Radiometer M295Ag Metal Electrode has a PTFE body and silver rod sensor. It is recommended for silver halide titrations. The M295Ag has a fixed 1 meter cable with banana plug.