Learn More
Electron Microscopy Sciences EMS M3 Series Trimmer Model M3-36
SDP
Supplier:  Electron Microscopy Sciences 7488536
Encompass_Preferred
Durable, accurate, and easy to use trimmer. Our cutting head is made from die-cast alloy construction, which adds strength, maintains alignment, and guarantees years of service. It's molded to the shape of your hand with an exclusive Comfort Grip. Within the housing is a hollow–ground tool-steel rotary blade that is self-sharpening for a long life of clean, precise cuts.
Dual steel rails guide the cutting head with complete stability and precision for accurate, repeatable cuts. Four nylon bearings in the head further ensure quiet, effortless cutting. The extruded aluminum supports on each end lock in alignment. A transparent paper guide automatically holds material in place without risk of damage. An extruded aluminum alignment scale is factory-locked to be perfectly square to the blade edge. The alignment scale has measurements in inches and centimeters and accepts the Slide Guide for making repeatable cuts.
Catalog No. 50-305-04