Learn More
Molecular Devices Molecular Devices Microplate Reader, Multi-Mode, Top/Bottom Read, 200-1000 nm
Catalog No. 5000014677
Encompass_Preferred
The SpectraMax M4 is a multi-mode microplate reader designed for absorbance, fluorescence, luminescence, and time-resolved fluorescence assays. It supports top and bottom reads and is engineered for flexible, routine laboratory measurements across a range of assay types.
  • Multi-mode detection: absorbance, fluorescence, luminescence, TRF.
  • Top and bottom reading for flexible assay configurations.
  • Absorbance range 200-1000 nm, fluorescence/luminescence/TRF 250-850 nm.
  • Dual-monochromator based detection for spectral flexibility.
  • Compatible with standard microplate formats, including 96-well plates.
  • Suitable for enzyme assays, nucleic acid and protein quantitation, and reporter assays.
Catalog No. 50-000-14677 Supplier Molecular Devices Supplier No. M4G
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M4 is a multi-mode microplate reader designed for absorbance, fluorescence, luminescence, and time-resolved fluorescence assays. It supports top and bottom reads and is engineered for flexible, routine laboratory measurements across a range of assay types.
  • Multi-mode detection: absorbance, fluorescence, luminescence, TRF.
  • Top and bottom reading for flexible assay configurations.
  • Absorbance range 200-1000 nm, fluorescence/luminescence/TRF 250-850 nm.
  • Dual-monochromator based detection for spectral flexibility.
  • Compatible with standard microplate formats, including 96-well plates.
  • Suitable for enzyme assays, nucleic acid and protein quantitation, and reporter assays.