Learn More
Diapharma Group, Inc. M30 Apoptosense CK18 Kit, 96 tests
SDP
Supplier:  Diapharma Group, Inc. P10011
Encompass
The M30 Apoptosense CK18 Kit (ELISA) is a unique tool for reliable and reproducible quantitative detection of apoptosis in epithelial keratin 18 (K18) (cytokeratin 18 (CK18)) positive cells. The specificity is reliable because the M30 Apoptosense ELISA solely quantifies the accumulation of soluble caspase-cleaved keratin 18, a product of apoptosis. The assay is intended for subject serum or plasma. Another useful application is the detection of tumor apoptosis in mouse xenograft models using plasma samples.
Catalog No. NC0676871