Learn More
Chemglass Life Sciences M30 Blind Stopper
SDP
Catalog No. 501532115
Encompass
M30 Blind Stopper. Stoppers can be used to close off any of the unused openings in the head plate. The M6, M10 and M12 sizes are fitted into the head plate from the inside, leaving a flat surface, while the M18, M27 and G3/4 are threaded into the ports from the outside. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.
Catalog No. 50-153-2115 Supplier Chemglass Life Sciences Supplier No. CLS138080
M30 Blind Stopper. Stoppers can be used to close off any of the unused openings in the head plate. The M6, M10 and M12 sizes are fitted into the head plate from the inside, leaving a flat surface, while the M18, M27 and G3/4 are threaded into the ports from the outside. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.