Learn More
Enzo Life Sciences M344 (1mg). CAS: 251456-60-7
SDP
Supplier:  Enzo Life Sciences ALX270297M001
Encompass_Preferred
HDAC inhibitor. Potent inhibitor of histone deacetylases (HDACs) (IC50≤1µM). Alternative name: 4-Dimethylamino-N-(6-hydroxycarbamoylhexyl)-benzamide. Formula: C16H25N3O3. MW: 307.4. Purity: ≥98% (HPLC). Appearance: White crystalline solid. Solubility: Soluble in DMSO (1mg/ml). Long Term Storage: +4°C. Handling: Protect from light.
Catalog No. 50-200-8476