Learn More
Selleck Chemical LLC M344-5mg
SDP
Catalog No. 502390077
Encompass_Preferred
M344 is a potent HDAC inhibitor with IC50 of 100 nM and able to induce cell differentiation.
Catalog No. 50-239-0077 Supplier Selleck Chemical LLC Supplier No. S27795MG
M344 is a potent HDAC inhibitor with IC50 of 100 nM and able to induce cell differentiation.