Learn More
Abcam M344, HDAC inhibitor, 1MG
Catalog No. 502706651
Encompass_Preferred
MW 307.39 Da, Purity >98%. HDAC inhibitor (IC₅₀ = 100 nM). Shows selectivity for HDAC6. Inhibits growth of various cancer cells. Apoptosis inducer. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6651 Supplier Abcam Supplier No. AB1414011MG
MW 307.39 Da, Purity >98%. HDAC inhibitor (IC₅₀ = 100 nM). Shows selectivity for HDAC6. Inhibits growth of various cancer cells. Apoptosis inducer. .
The product is subject to the following: Abcam Restricted Use Statement