Learn More
CPC Scientific H-Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GALA004A
Encompass
Galanin antagonist that binds with high specificity and affinity to galanin binding sites and provides a new principle in the treatment of dementia and related disorders. At low concentrations, this chimeric peptide antagonizes the effect of galanin, whereas at concentrations above 10nM M35 it acts as a galanin receptor agonist.ONE-LETTER SEQUENCE: GWTLNSAGYLLGPPPGFSPFR-NH2MOLECULAR FORMULA: C107H153N27O26MOLECULAR WEIGHT:2233.56STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [142846-71-7], SYNONYMS: Galanin (1-13)-Bradykinin (2-9) amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES: K. Kask et al., Regulatory Peptide, 59, 341 (1995)
Catalog No. 50-210-9310