Learn More
MARSHALL SCIENTIFIC Branson M3800 Mechanical Ultrasonic Cleaner
SDP
Catalog No. NC2590773
Encompass_Preferred
Bransonic M and MH Series cleaners are known for offering a level of precision and reliability you have come to expect from Branson. M Series includes 60-minute mechanical timer and continuous operation. MH Series includes 60-minute mechanical timer and continuous operation with heating capability. Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance, even with multiple users or when bath conditions change.
Catalog No. NC2590773 Supplier MARSHALL SCIENTIFIC Supplier No. CPX952316R
Bransonic M and MH Series cleaners are known for offering a level of precision and reliability you have come to expect from Branson. M Series includes 60-minute mechanical timer and continuous operation. MH Series includes 60-minute mechanical timer and continuous operation with heating capability. Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance, even with multiple users or when bath conditions change.