Learn More
Medchemexpress LLC M4 mAChR agonist-1 | 785705-53-5 | 99.0% | 290.38 g·mol⁻¹ | C14H18N4OS | 5 MG
SDP
Catalog No. 5000222127
Encompass_Preferred
M4 mAChR agonist-1 is a small-molecule research compound (compound 10a) that acts as an agonist of the human M4 muscarinic acetylcholine receptor. The compound has CAS 785705-53-5, formula C14H18N4OS, and molecular weight 290.38 g·mol⁻¹. Reported activity for human M4 is EC50 >10 μM. Supplied at high purity for in vitro pharmacology and receptor research; for research use only.
  • Small-molecule M4 muscarinic receptor agonist.
  • Reported EC50 >10 μM for human M4.
  • High purity suitable for biochemical assays.
  • Available in multiple mg-scale quantities.
  • Acceptable for in vitro pharmacology and receptor studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-22127 Supplier Medchemexpress LLC Supplier No. HY1470285MG
M4 mAChR agonist-1 is a small-molecule research compound (compound 10a) that acts as an agonist of the human M4 muscarinic acetylcholine receptor. The compound has CAS 785705-53-5, formula C14H18N4OS, and molecular weight 290.38 g·mol⁻¹. Reported activity for human M4 is EC50 >10 μM. Supplied at high purity for in vitro pharmacology and receptor research; for research use only.
  • Small-molecule M4 muscarinic receptor agonist.
  • Reported EC50 >10 μM for human M4.
  • High purity suitable for biochemical assays.
  • Available in multiple mg-scale quantities.
  • Acceptable for in vitro pharmacology and receptor studies.
  • For research use only; not for human or clinical use.