Learn More
Medchemexpress LLC M40 TFA | 143896-17-7 | 98.3% | 1980.31 | 5 MG
SDP
Catalog No. 5000425165
Encompass_Preferred
M40 TFA is the TFA salt form of M40, functioning as an antagonist or weak agonist for galanin receptors. Its activity varies depending on specific galanin receptor subtypes found in regions like the brain, hypothalamus, hippocampus, amygdala, and pancreas. It has demonstrated efficacy in various in vivo studies.
  • Blocks central cardiovascular responses induced by specific galanin fragments in rat models.
  • Acts as an antagonist against galanin on memory tasks in rat models.
  • Shows potential in mitigating cognitive deficits related to Alzheimer's disease.
Catalog No. 50-004-25165 Supplier Medchemexpress LLC Supplier No. HYP1025A5MG
M40 TFA is the TFA salt form of M40, functioning as an antagonist or weak agonist for galanin receptors. Its activity varies depending on specific galanin receptor subtypes found in regions like the brain, hypothalamus, hippocampus, amygdala, and pancreas. It has demonstrated efficacy in various in vivo studies.
  • Blocks central cardiovascular responses induced by specific galanin fragments in rat models.
  • Acts as an antagonist against galanin on memory tasks in rat models.
  • Shows potential in mitigating cognitive deficits related to Alzheimer's disease.