Learn More
Medchemexpress LLC M40 (TFA) | 98.3% | 1980.31 (free base) | 1 MG
SDP
Catalog No. 5000425506
Encompass_Preferred
This product is the TFA salt form of the compound M40. It functions as either an antagonist or a weak agonist for the galanin receptor, with its specific action depending on the particular subtype of galanin receptor present in different brain regions, including the hypothalamus, hippocampus, amygdala, and pancreas.
  • Exhibits antagonist efficacy against galanin on memory tasks in models.
  • Demonstrates potency in alleviating cognitive deficits associated with Alzheimer's disease.
  • Blocks central cardiovascular responses induced by specific galanin fragments.
  • Reduced vasopressor response and heart rate induced by galanin-(1-15).
  • Blocks impairment in choice accuracy in animal models.
Catalog No. 50-004-25506 Supplier Medchemexpress LLC Supplier No. HYP1025A1MG
This product is the TFA salt form of the compound M40. It functions as either an antagonist or a weak agonist for the galanin receptor, with its specific action depending on the particular subtype of galanin receptor present in different brain regions, including the hypothalamus, hippocampus, amygdala, and pancreas.
  • Exhibits antagonist efficacy against galanin on memory tasks in models.
  • Demonstrates potency in alleviating cognitive deficits associated with Alzheimer's disease.
  • Blocks central cardiovascular responses induced by specific galanin fragments.
  • Reduced vasopressor response and heart rate induced by galanin-(1-15).
  • Blocks impairment in choice accuracy in animal models.