Learn More
Medchemexpress LLC M40 (TFA) | 98.3% | 1980.31 (free base) | 10 MG
SDP
Catalog No. 5000425440
Encompass_Preferred
M40 TFA is the trifluoroacetate salt form of M40, functioning as an antagonist or weak agonist for galanin receptor subtypes across various brain regions including the hypothalamus, hippocampus, amygdala, and pancreas.
  • Modulates galanin receptor activity
  • Blocks central cardiovascular responses induced by galanin-(1-15)
  • Demonstrates antagonist efficacy on memory tasks
  • May alleviate cognitive deficits associated with Alzheimer's disease
  • White to off-white solid appearance
  • Recommended storage: -80°C for 2 years or -20°C for 1 year as powder; -80°C for 6 months or -20°C for 1 month in solvent (sealed, away from moisture)
Catalog No. 50-004-25440 Supplier Medchemexpress LLC Supplier No. HYP1025A10MG
M40 TFA is the trifluoroacetate salt form of M40, functioning as an antagonist or weak agonist for galanin receptor subtypes across various brain regions including the hypothalamus, hippocampus, amygdala, and pancreas.
  • Modulates galanin receptor activity
  • Blocks central cardiovascular responses induced by galanin-(1-15)
  • Demonstrates antagonist efficacy on memory tasks
  • May alleviate cognitive deficits associated with Alzheimer's disease
  • White to off-white solid appearance
  • Recommended storage: -80°C for 2 years or -20°C for 1 year as powder; -80°C for 6 months or -20°C for 1 month in solvent (sealed, away from moisture)