Learn More
Selleck Chemical LLC Lartesertib-E1057-5MG
SDP
Catalog No. 502389780
Encompass_Preferred
Lartesertib inhibits Ataxia telangiectasia-mutated (ATM) kinase activity with a sub-nanomolar potency and shows remarkable selectivity against other protein kinases suppresses DSB repair clonogenic cancer cell growth and potentiates antitumor activity of ionizing radiation in cancer cell lines
Catalog No. 50-238-9780 Supplier Selleck Chemical LLC Supplier No. E10575MG
Lartesertib inhibits Ataxia telangiectasia-mutated (ATM) kinase activity with a sub-nanomolar potency and shows remarkable selectivity against other protein kinases suppresses DSB repair clonogenic cancer cell growth and potentiates antitumor activity of ionizing radiation in cancer cell lines