Learn More
Medchemexpress LLC HY-141891 5mg , M435-1279 CAS:1359431-16-5 Purity:>98%
SDP
Catalog No. 502261845
Encompass_Preferred
HY-141891 5mg M435-1279 CAS: 1359431-16-5 M435-1279 is a UBE2T inhibitor. M435-1279 inhibits the Wnt/β-catenin signaling pathway hyperactivation through blocking UBE2T-mediated degradation of RACK1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1845 Supplier Medchemexpress LLC Supplier No. HY1418915MG
Add to Cart
Add to Cart
HY-141891 5mg M435-1279 CAS: 1359431-16-5 M435-1279 is a UBE2T inhibitor. M435-1279 inhibits the Wnt/β-catenin signaling pathway hyperactivation through blocking UBE2T-mediated degradation of RACK1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.