Learn More
Medchemexpress LLC HY-112274 5mg Medchemexpress, M443 CAS:1820684-31-8 Purity:>98%
SDP
Catalog No. 501969070
Encompass_Preferred
Medchemexpress, HY-112274 5mg M443 CAS:1820684-31-8 M443 is an irreversible and specific inhibitor of MRK, with an IC50<125 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9070 Supplier Medchemexpress LLC Supplier No. HY1122745MG
Medchemexpress, HY-112274 5mg M443 CAS:1820684-31-8 M443 is an irreversible and specific inhibitor of MRK, with an IC50<125 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.