Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M4K2163 dihydrochloride | 2863635-04-3 | 98.3% | 480.40 | C24H28Cl2FN3O2 | 10 MG
SDP

Catalog No. 5000287763
Encompass_Preferred

M4K2163 dihydrochloride is a potent, selective activin receptor-like kinase-2 (ALK2) inhibitor with blood-brain barrier permeability intended for preclinical CNS research. It displays an IC50 of 19 nM and is supplied as a high-purity solid for laboratory use.

  • Potent, selective ALK2 inhibitor (IC50 = 19 nM).
  • Blood-brain barrier permeable, suitable for central nervous system research.
  • High purity (98.3%) and characterized solid form (white to yellow).
  • Molecular weight 480.40; formula C24H28Cl2FN3O2.
  • Storage: sealed at -20°C; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Available in 10 mg vial for laboratory use.

Catalog No. 50-002-87763 Supplier Medchemexpress LLC Supplier No. HY15593910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M4K2163 dihydrochloride is a potent, selective activin receptor-like kinase-2 (ALK2) inhibitor with blood-brain barrier permeability intended for preclinical CNS research. It displays an IC50 of 19 nM and is supplied as a high-purity solid for laboratory use.

  • Potent, selective ALK2 inhibitor (IC50 = 19 nM).
  • Blood-brain barrier permeable, suitable for central nervous system research.
  • High purity (98.3%) and characterized solid form (white to yellow).
  • Molecular weight 480.40; formula C24H28Cl2FN3O2.
  • Storage: sealed at -20°C; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Available in 10 mg vial for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.