Learn More
VEOLIA WTS M500/500 SDBS SUITABILITY SET
Catalog No. NC2894274
Encompass
M500/500 SDBS Suitability SetSuitability standard set to meet Japanese and KoreanPharmacopeia requirements for Model M500 and 500 RL TOC Analyzers.This standard set contains two 30 mL DUCT vial standards: a Reagent WaterBlank and a 500 µg C/L as SDBS standard.
Catalog No. NC2894274 Supplier VEOLIA WTS Supplier No. STD 7442901
M500/500 SDBS Suitability SetSuitability standard set to meet Japanese and KoreanPharmacopeia requirements for Model M500 and 500 RL TOC Analyzers.This standard set contains two 30 mL DUCT vial standards: a Reagent WaterBlank and a 500 µg C/L as SDBS standard.