Learn More
IDEX Health & Science Upchurch M6 FEMALE TO FEMALE LUER
Catalog No. NC1919479
Encompass_Preferred
Idex Threaded Luer PEEK Adapter, Black, For Use With: To Female M6, Luer Adapter, Diameter Inner: 0.05 in., Temperature Max: 125 deg.C
Catalog No. NC1919479 Supplier IDEX Health & Science Upchurch Supplier No. P660
Idex Threaded Luer PEEK Adapter, Black, For Use With: To Female M6, Luer Adapter, Diameter Inner: 0.05 in., Temperature Max: 125 deg.C