Learn More
IDEX Health & Science Upchurch M6 MALE TO FEMALE LUER TEFZEL
Catalog No. NC1919832
Encompass_Preferred
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.05 in., Black, For Use With: To Female M6, Luer Adapter, ETFE, Temperature Max: 80 deg.C
Catalog No. NC1919832 Supplier IDEX Health & Science Upchurch Supplier No. P686
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.05 in., Black, For Use With: To Female M6, Luer Adapter, ETFE, Temperature Max: 80 deg.C