Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady M610 Handheld Label Maker with Hard Case

Catalog No. 17100918 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Gray, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-918 Gray, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-918 Supplier Brady Supplier No. M610KITTAA
Add to Cart
Add to Cart

Get the durability on your work demands, military-grade shock resistance, below freezing operating temps and the widest range of data entry options

  • Kit includes a hard case
  • TAA compliant for US government contracts
  • Design and print labels using a keypad or desktop software
  • Work longer with an enhanced Gen3 Li-iON battery-print 4500 labels on a full charge
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 2 per second print speed
  • Gain added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 6 drops
  • Operate your printer in 0° to 115°F temperatures
  • NOTICE- Brady North America is not authorized to ship this item via air or to Alaska or Hawaii

Specifications

Product Type Label Printer Kit
Color Gray, Yellow
Display Type Color LCD
For Use With (Equipment) BM Series Printers
Material Plastic
Print Technology Thermal Transfer
Length (English) 4.19 in.
Width (English) 4.22 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.