Learn More
Medchemexpress LLC M617 TFA | 99.4% | 2475.67 | 5 MG

Supplier: Medchemexpress LLC HYP1131A5MG
M617 TFA is a selective galanin receptor 1 (GAL1) agonist that promotes GLUT4 expression and enhances GLUT4 content in cardiac muscle of type 2 diabetic rats. It has a high selectivity with Kis of 0.23 nM for GAL1 and 5.71 nM for GAL2.
- Selective galanin receptor 1 (GAL1) agonist
- Promotes GLUT4 expression
- Enhances GLUT4 content in cardiac muscle
- Molecular weight of 2475.67
- Molecular formula C114H162F3N29O30
- Purity of 99.38% (LCMS)
- White to off-white solid appearance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.