Learn More
Bachem M65 Trifluoroacetate
SDP
Supplier:  Bachem 4067721.0500
Encompass_Preferred
M65 Trifluoroacetate, H-Cys-Asp-Ala-Thr-Cys-Gln-Phe-Arg-Lys-Ala-Ile-Asp-Asp-Cys-Gln-Lys-Gln-Ala-His-His-Ser-Asn-Val-Pro-Gly-Asn-Ser-Val-Phe-Lys-Glu-Cys-Met-Lys-Gln-Lys-Lys-Lys-Glu-Phe-Lys-Ala-NH2, 1872440-65-7, C205H326N64O61S5, Mr 4823.57, 0.5mg. PAC1 Receptor Antagonist M65. Potent and specific PAC1 receptor antagonist.
Catalog No. 50-260-123