Learn More
CPC Scientific H-Cys-Asp-Ala-Thr-Cys-Gln-Phe-Arg-Lys-Ala-Ile-Asp-Asp-Cys-Gln-Lys-Gln-Ala-His-His-Ser-Asn-Val-Pro-Gly-Asn-Ser-Val-Phe-Lys-Glu-Cys-Met-Lys-Gln-Lys-Lys-Glu-Phe-Lys-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PACR001B
Encompass
PAC1 receptor antagonistONE-LETTER SEQUENCE: CDATCQFRKAIDDCQKQAHHSNVPGNSVFKECMKQKKEFKA-NH2 (Cys1 & Cys5, Cys14 & Cys32 bridge)MOLECULAR FORMULA: C199H314N62O60S5MOLECULAR WEIGHT:4695.39STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: O.Moro et al., J. Biol. Chem., 274, 23103 (1999); E.A.Lerner et al., Peptides, 28, 1651 (2007)
Catalog No. 50-211-0069