Learn More
Medchemexpress LLC M65 TFA | 99.98% | 4823.53 (free base) | 5 MG
SDP
Catalog No. 5000399508
Encompass_Preferred
M65 TFA is a deleted peptide of maxadilan (61 a.a.), specifically missing residues between positions 24 and 42. It functions as a specific antagonist of the PACAP type 1 receptor, inhibiting ANP secretion, and is utilized in relevant research.
  • Deleted peptide of maxadilan (61 a.a.) with a deletion of residues between positions 24 and 42.
  • Acts as a specific antagonist of the PACAP type 1 receptor.
  • Inhibits ANP secretion.
  • Completely blocks cAMP accumulation stimulated by 100 nM of VIP in rat cortical neurons at 1 μM concentration.
  • Partially inhibits cAMP accumulation stimulated by 1 nM of maxadilan in rat cortical neurons at 1 μM concentration.
Catalog No. 50-003-99508 Supplier Medchemexpress LLC Supplier No. HYP4127A5MG
M65 TFA is a deleted peptide of maxadilan (61 a.a.), specifically missing residues between positions 24 and 42. It functions as a specific antagonist of the PACAP type 1 receptor, inhibiting ANP secretion, and is utilized in relevant research.
  • Deleted peptide of maxadilan (61 a.a.) with a deletion of residues between positions 24 and 42.
  • Acts as a specific antagonist of the PACAP type 1 receptor.
  • Inhibits ANP secretion.
  • Completely blocks cAMP accumulation stimulated by 100 nM of VIP in rat cortical neurons at 1 μM concentration.
  • Partially inhibits cAMP accumulation stimulated by 1 nM of maxadilan in rat cortical neurons at 1 μM concentration.