Learn More
Aviva Systems Biology Corp M6PR Antibody - N-terminal region (ARP43519_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43519_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against M6PR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human M6PR. Alternate names: CD-MPR, FLJ32994, MPR46, SMPR, MPR 46, MPR-46ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53367