Learn More
MSC Value Collection MASC0060025CP M6x1.00, 25mm Long, Metric Coarse Hex Socket Cap Screw 18-8 Austenitic Grade A2 Stainless Steel, Partially Threaded
Catalog No. NC1913517
Encompass
Value Collection MASC0060025CP M6x1.00, 25mm Long, Metric Coarse Hex Socket Cap Screw 18-8 Austenitic Grade A2 Stainless Steel, Partially Threaded
Catalog No. NC1913517 Supplier MSC Supplier No. 68016047
Value Collection MASC0060025CP M6x1.00, 25mm Long, Metric Coarse Hex Socket Cap Screw 18-8 Austenitic Grade A2 Stainless Steel, Partially Threaded