Learn More
Medchemexpress LLC M7(3'ma-biotin)gppp(2'ome)apg ammonium solution (25mm) | 90.0% | 1385.00 (free base) | 100 μL
SDP
Catalog No. 5000391543
Encompass_Preferred
m7(3'Ma-Biotin)Gppp(2'OMe)ApG ammonium is a Biotin-labeled cap analogue that can be used for mRNA synthesis. The efficiency of mRNA delivery can be determined by the detection of Biotin by flow cytometry, which can be used to track mRNA in cells and in living animals and to screen delivery lipid ratios.
  • Biotin-labeled cap analogue for mRNA synthesis.
  • Can be used to determine the efficiency of mRNA delivery by detecting Biotin via flow cytometry.
  • Useful for tracking mRNA in cells and living animals.
  • Can be used to screen delivery lipid ratios.
Catalog No. 50-003-91543 Supplier Medchemexpress LLC Supplier No. HY1641903.3MG
m7(3'Ma-Biotin)Gppp(2'OMe)ApG ammonium is a Biotin-labeled cap analogue that can be used for mRNA synthesis. The efficiency of mRNA delivery can be determined by the detection of Biotin by flow cytometry, which can be used to track mRNA in cells and in living animals and to screen delivery lipid ratios.
  • Biotin-labeled cap analogue for mRNA synthesis.
  • Can be used to determine the efficiency of mRNA delivery by detecting Biotin via flow cytometry.
  • Useful for tracking mRNA in cells and living animals.
  • Can be used to screen delivery lipid ratios.