Learn More
Brady™ M710 Label Printer AC Adapter/Charger
Catalog No. 17986735 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Brady Label Printers
Catalog No. For Use With (Equipment)
17-986-735 Brady Label Printers
1 options
Catalog No. 17-986-735 Supplier Brady™ Supplier No. M710AC
Only null left
Add to Cart
Add to Cart
Get the power you need to work all day with this AC adapter/charger.
  • AC Adapter powers the M710 Label Printer and charges the battery pack.
  • Use the Brady approved power supply specifically selected for the M710 Label Printer.
  • Power supply voltage 100 to 240 V, output 5.0 A, 90.0 W.
Specifications
Product Type AC Adapter
For Use With (Equipment) Brady Label Printers
Model M710