Learn More
Medchemexpress LLC M77976 10mM 1mL | 394237-61-7 | 1 ML
SDP
Catalog No. 5000229609
Encompass_Preferred
M77976 is a small-molecule, ATP-competitive inhibitor of pyruvate dehydrogenase kinase 4 (PDK4) supplied as a 10 mM solution in DMSO (1 mL) for research use. It shows enzymatic activity (IC50 648 μM) and cellular activity (HCT-116 GI50 33.3 μM), with chemical formula C17H16N2O3 and molecular weight 296.32.
  • Specific ATP-competitive inhibition of PDK4 (IC50 648 μM).
  • Measurable cellular activity in HCT-116 cells (GI50 33.3 μM).
  • High purity suitable for research (99.4%).
  • Provided as a 10 mM solution in DMSO, 1 mL volume.
  • High solubility in DMSO (250 mg/mL) and compatible with common in vivo vehicles.
  • Stable when stored under recommended conditions for powder and solution.
Catalog No. 50-002-29609 Supplier Medchemexpress LLC Supplier No. HY11470210MM 1ML
M77976 is a small-molecule, ATP-competitive inhibitor of pyruvate dehydrogenase kinase 4 (PDK4) supplied as a 10 mM solution in DMSO (1 mL) for research use. It shows enzymatic activity (IC50 648 μM) and cellular activity (HCT-116 GI50 33.3 μM), with chemical formula C17H16N2O3 and molecular weight 296.32.
  • Specific ATP-competitive inhibition of PDK4 (IC50 648 μM).
  • Measurable cellular activity in HCT-116 cells (GI50 33.3 μM).
  • High purity suitable for research (99.4%).
  • Provided as a 10 mM solution in DMSO, 1 mL volume.
  • High solubility in DMSO (250 mg/mL) and compatible with common in vivo vehicles.
  • Stable when stored under recommended conditions for powder and solution.