Learn More
New England Biolabs, Inc. m7G(5')ppp(5')G RNA Cap Structure Analog – 1 umol
SDP
Supplier:  New England Biolabs, Inc. S1404S
Encompass
For most RNAs, elimination of the cap structure causes a loss of stability, especially against exonuclease degradation, and a decrease in the formation of the initiation complex of mRNAs for protein synthesis. Certain prokaryotic mRNAs containing a 5' terminal cap structure are translated as efficiently as or more efficiently than eukaryotic mRNAs in a eukaryotic cell-free protein synthesizing system. Also a cap requirement has been observed for splicing eukaryotic substrate RNAs.
Catalog No. 50-812-652