Learn More
Apexbio Technology LLC m7GTP(Sodium salt) 104809-18-9 50ul (100 mM)
SDP
Catalog No. 5000064067
Encompass_Preferred
m7GTP (Sodium salt) chemically known as 7-methylguanosine 5 -triphosphate structurally resembles the 5 -cap of eukaryotic messenger RNA (mRNA) Due to its affinity for the eukaryotic initiation factor 4E (eIF4E) this compound frequently functions as an affinity ligand for isolating and studying translation initiation complexes Mechanistically m7GTP binds specifically to eIF4E thereby competitively inhibiting interaction with native capped mRNA and disrupting subsequent initiation complex formation during in vitro protein synthesis Consequently it serves as an essential research tool for examining translation initiation processes delineating mRNA cap recognition pathways and characterizing regulatory mechanisms governing eukaryotic protein synthesis
Catalog No. 50-000-64067 Supplier Apexbio Technology LLC Supplier No. B88010.05
m7GTP (Sodium salt) chemically known as 7-methylguanosine 5 -triphosphate structurally resembles the 5 -cap of eukaryotic messenger RNA (mRNA) Due to its affinity for the eukaryotic initiation factor 4E (eIF4E) this compound frequently functions as an affinity ligand for isolating and studying translation initiation complexes Mechanistically m7GTP binds specifically to eIF4E thereby competitively inhibiting interaction with native capped mRNA and disrupting subsequent initiation complex formation during in vitro protein synthesis Consequently it serves as an essential research tool for examining translation initiation processes delineating mRNA cap recognition pathways and characterizing regulatory mechanisms governing eukaryotic protein synthesis