Learn More
Cayman Chemical M8-B (hydrochloride), Purity: ≥98%, CAS: 883976-12-3, 10 mg
SDP
Catalog No. 502986333
Encompass_Preferred
A selective antagonist of TRPM8 that blocks activation by cold, icilin, or menthol in vitro (IC50s = 7.8, 26.9, and 64.3 nM, respectively); decreases deep body temperature in wild-type mice and rats but has no effect on body temperature in TRPM8 knockout mice
Catalog No. 50-298-6333 Supplier Cayman Chemical Supplier No. 2164610 MG
A selective antagonist of TRPM8 that blocks activation by cold, icilin, or menthol in vitro (IC50s = 7.8, 26.9, and 64.3 nM, respectively); decreases deep body temperature in wild-type mice and rats but has no effect on body temperature in TRPM8 knockout mice