Learn More
Leica Microsystems M80 Stereo Microscope
Catalog No. 01672669 Shop All Leica Microsystems Products
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
01-672-669 Binocular
1 options
Catalog No. 01-672-669 Supplier Leica Microsystems Supplier No. 8122687
Add to Cart
Add to Cart
Request a Quote
The Leica M80 stereo microscope: ergonomic, precise, and ideal for professionals. With superior optics and user-friendly features, it ensures efficiency and comfort during prolonged usage. Elevate your inspection experience with the Leica M80, where precision meets ergonomic design.
The Leica M80 stereo microscope sets the standard for precision inspection with its ergonomic design. Crafted for professionals, this microscope offers superior optics and a comfortable ergonomic setup, ensuring prolonged usage without compromising efficiency. The Leica M80 seamlessly integrates clarity and user-friendly features, making it the ideal choice for professionals seeking an ergonomic solution for detailed inspection tasks. Elevate your inspection experience with the Leica M80, where precision meets ergonomic design for optimal performance in the field of microscopy.
  • Magnification range: 7.5x -60x (1x objective, 10x eyepieces)
  • Optics parfocal
  • 8 click stop zoom settings
  • Modular system
  • Suitable for achromatic, plan or planapo objectives
  • Objective thread M60
  • ESD
Includes:
  • 10450155 Leica M80 optics carrier
  • 10450910 Eyepiece 10x/23B, adjustable, 3d gen x2
  • 10450158 Ergo Tube 10°-50°, binocular
  • 10450159 Objective achromat 1.0x, WD = 90 mm
  • 10445661 Power cable, 2m, USA
  • 10450172 Focus drive 500 mm M-Series Routine
  • 10450173 Microscope carrier
  • 10450049 Inc. light base, large w. AntiShock feet
  • 10450434 Intermediate adapter for TL and IL Base
  • 10450271 Leica LED3000 RL, 58 mm
  • 10450337 Polarization set for LED3000 RL
  • 10450266 Power Supply for LED3000/LED5000
  • 10447039 Dust cover antistatic, small
TRUSTED_SUSTAINABILITY
Specifications
Voltage 100/240 V
Magnification Power 7.5X to 60X
Objective Types Achromatic
Product Type Educational Microscope
Head Style Binocular
Lighting Type LED, Reflected Illumination
Eyepiece WF 10X/23 mm, Adjustable