Learn More
Abcam M871, Galanin GAL2 receptor antagonist, 1MG
Supplier:  Abcam AB1411591MG
Encompass_Preferred
MW 2287.64 g/mol. Selective galanin GAL₂ receptor antagonist (Ki values are 13.1 and 420 nM at GAL₂ and GAL₁ receptors, respectively). Blocks pronociceptive effects of GAL₂ agonists including AR-M1896. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6846