Learn More
MARSHALL SCIENTIFIC Branson M8800 Mechanical Ultrasonic Cleaner
SDP
Supplier:  MARSHALL SCIENTIFIC CPX952816R
Encompass_Preferred
Bransonic M and MH Series cleaners are known for offering a level of precision and reliability you have come to expect from Branson. M Series includes 60-minute mechanical timer and continuous operation. MH Series includes 60-minute mechanical timer and continuous operation with heating capability. Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance, even with multiple users or when bath conditions change.
Catalog No. NC2590786