Learn More
Medchemexpress LLC HY-133016 5mg , M8891 CAS:1464842-09-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1330165MG
Encompass_Preferred
HY-133016 5mg M8891 CAS: 1464842-09-8 M8891 is an orally active, reversible and brain penetrant Methionine Aminopeptidase-2 (MetAP-2) inhibitor with an IC50 of 54 nM and a Ki of 4.33 nM. M8891 does not inhibit MetAP-1 (IC50>10 µM). M8891 inhibits growth of primary endothelial cells as well as tumor cells and demonstrates antiangiogenic and antitumoral activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0738