Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc M9 CA Medium, pH 7.1, 500 mL
SDP

Catalog No. 502209767
Encompass_Preferred

M9 CA Media contains the essentials for bacterial species to grow. M9 Medium is used for the following applications: propagation of E. coli strains, maintaining positive selective pressure on recombinant strains of E. coli with plasmids coding for the ability to produce essential substances such as amino acids or vitamin, and maintaining stocks of F’-containing bacteria for use with M13 bacteriophage.

Without Phenol Red
Glucose as a carbon source
Casamino acids as a nitrogen source

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9767 Supplier Quality Biological Inc Supplier No. 340017101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M9 CA Media contains the essentials for bacterial species to grow. M9 Medium is used for the following applications: propagation of E. coli strains, maintaining positive selective pressure on recombinant strains of E. coli with plasmids coding for the ability to produce essential substances such as amino acids or vitamin, and maintaining stocks of F’-containing bacteria for use with M13 bacteriophage.

Without Phenol Red
Glucose as a carbon source
Casamino acids as a nitrogen source

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.