Learn More
Bioworld M9 Medium (5x), Sterile, 250 mL
Catalog No. 50254439
Encompass_Preferred
M9 Medium (5x)Media for the cultivation of recombinant E. coli and other Auxotrophs Biotechnology Grade 5x Sterile Liquid Concentrate
Catalog No. 50-254-439 Supplier Bioworld Supplier No. 306210182
M9 Medium (5x)Media for the cultivation of recombinant E. coli and other Auxotrophs Biotechnology Grade 5x Sterile Liquid Concentrate