Learn More
Bioworld M9 Minimal Salts Broth, Sterile, 1 L
Catalog No. 50254441
Encompass_Preferred
M9 Minimal Salt BrothFor the cultivation of recombinant strains of E. coli May also be used for the maintenance of F'-containing strains of E. coli with M13 single-stranded, filamentous bacteriophage Contains:1X M9 Salts (Na2HPO4, KH2PO4, NaCl, NH4Cl) Magnesium Sulfate Calcium ChlorideGlucose
Catalog No. 50-254-441 Supplier Bioworld Supplier No. 306210191
Add to Cart
Add to Cart
M9 Minimal Salt BrothFor the cultivation of recombinant strains of E. coli May also be used for the maintenance of F'-containing strains of E. coli with M13 single-stranded, filamentous bacteriophage Contains:1X M9 Salts (Na2HPO4, KH2PO4, NaCl, NH4Cl) Magnesium Sulfate Calcium ChlorideGlucose