Learn More
Bioworld M9 Minimal Salts (5x), Dry Powder, 1 kg
Catalog No. 50254436
Encompass_Preferred
M9 Minimal Salts (5x)Salts used to make M9 Minimal MediumFor cultivation of recombinant strains of E. coli May also be used for maintenance of F'-containing strains of E. coli with M13 single-stranded, filamentous bacteriophagepH: 6.8Prepared Appearance: colorlessSoluble in waterContains:Sodium Phosphate, Dibasic Potassium Phosphate, Monobasic Sodium Chloride Ammonium Chloride
Catalog No. 50-254-436 Supplier Bioworld Supplier No. 306210152
M9 Minimal Salts (5x)Salts used to make M9 Minimal MediumFor cultivation of recombinant strains of E. coli May also be used for maintenance of F'-containing strains of E. coli with M13 single-stranded, filamentous bacteriophagepH: 6.8Prepared Appearance: colorlessSoluble in waterContains:Sodium Phosphate, Dibasic Potassium Phosphate, Monobasic Sodium Chloride Ammonium Chloride